Learn More
Advanced Materials Technology HALO 90 PFAS 2.7 2.1 X
Catalog No. 502556259
Encompass_Preferred
HALO PFAS is designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, along with EPA methodology requirements in mind, HALO PFAS offers a holistic solution.
Catalog No. 50-255-6259 Supplier Advanced Materials Technology Supplier No. 92812913
HALO PFAS is designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, along with EPA methodology requirements in mind, HALO PFAS offers a holistic solution.