Learn More
Advanced Materials Technology HALO 90 PFAS 2.7 2.1 X
Supplier: Advanced Materials Technology 92812613
HALO PFAS is designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, along with EPA methodology requirements in mind, HALO PFAS offers a holistic solution.