Learn More
Advanced Materials Technology HALO 90 PFP 2 3.0 X 250
Catalog No. 502556590
Encompass_Preferred
HALO PFP columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6590 Supplier Advanced Materials Technology Supplier No. 91813909
HALO PFP columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.