Learn More
Advanced Materials Technology HALO 90 PFP 2.7 3.0 X 3
Catalog No. 502556233
Encompass_Preferred
HALO PFP columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6233 Supplier Advanced Materials Technology Supplier No. 92813309
HALO PFP columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.