Learn More
Advanced Materials Technology HALO 90 PHENYL-HEXYL 2.7
Supplier: Advanced Materials Technology 98216706
HALO Phenyl-Hexyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Phenyl-Hexyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.