Learn More
Advanced Materials Technology HALO 90 RP-AMIDE 2.7 0.
Supplier:  Advanced Materials Technology 98216607
Encompass_Preferred
HALO RP-Amide columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-5593