Learn More
Advanced Materials Technology HALO 90 RP-AMIDE 5 10.0
Supplier: Advanced Materials Technology 95810407
HALO RP-Amide columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.