Learn More
Advanced Materials Technology HALO 90 Å PCS Phenyl-Hexyl, 2.7um, 3.0 mm x 100 mm
Catalog No. NC3203182
Encompass_Preferred
Built upon proven Fused-Core technology for speed and efficiency, the HALO PCS column products are positively charged surface chemistries designed to deliver improved peak shapes for basic compounds observed with standard Phenyl-Hexyl chemistry. Ideal for use with low ionic strength mobile phases, HALO PCS maintains peak symmetry at higher loading capacities and provides alternate selectivities from Phenyl-Hexyl bonded phases. The columns are optimized to deliver performance for reproducible, high efficiency LC and LCMS separations.
Catalog No. NC3203182 Supplier Advanced Materials Technology Supplier No. 92813618
Built upon proven Fused-Core technology for speed and efficiency, the HALO PCS column products are positively charged surface chemistries designed to deliver improved peak shapes for basic compounds observed with standard Phenyl-Hexyl chemistry. Ideal for use with low ionic strength mobile phases, HALO PCS maintains peak symmetry at higher loading capacities and provides alternate selectivities from Phenyl-Hexyl bonded phases. The columns are optimized to deliver performance for reproducible, high efficiency LC and LCMS separations.