Learn More
Advanced Materials Technology HALO 90 Å Biphenyl, 2.7 µm, 3.0 x 100 mm
Catalog No. NC2144774
Encompass_Preferred
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
This item is not returnable. View return policy
Catalog No. NC2144774 Supplier Advanced Materials Technology Supplier No. 92813611
HALO Biphenyl columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å Biphenyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.