Learn More
Advanced Materials Technology HALO 90 A C18, 2.7 µm, 1.5 x 50 mm
Supplier:  Advanced Materials Technology 9281X402
Encompass_Preferred
The New HALO 1.5 – Defining a New Dimension in Chromatography Delivering More PerformanceMore Sensitivity from conventional UHPLC SystemsHigher Ionization Efficiencies from LCMS systemsReduce Solvent Consumption compared to 2.1 mm ID columnsEasy to Implement microflow solution
Catalog No. NC2096806