Learn More
Advanced Materials Technology HALO 90 Å C18, 2 µm, 2.1 x 75 mm
Supplier:  Advanced Materials Technology 91812502
Encompass_Preferred
HALO C18 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å C18 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. NC1922494