Learn More
Sigma Aldrich Fine Chemicals Biosciences Halocarbon oil 700, 9002-83-9, MFCD00147324, 50 mL
Catalog No. NC0852076
Encompass
Halocarbon oil 700 has been used: to facilitate piercing of vitelline membranes with a glass capillary to make small holes on spider embryos, in microinjection procedure to track the vesicular stomatitis virus infection in Caenorhabditis elegans, and for live imaging of embryo sample using Zeiss LSM710 confocal system.
Catalog No. NC0852076 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H889850ML
Halocarbon oil 700 has been used: to facilitate piercing of vitelline membranes with a glass capillary to make small holes on spider embryos, in microinjection procedure to track the vesicular stomatitis virus infection in Caenorhabditis elegans, and for live imaging of embryo sample using Zeiss LSM710 confocal system.