Learn More
Advanced Materials Technology HALO 90 Å HILIC, 2.7 µm, 2.1 x 100 mm
Catalog No. 5090002110
Encompass_Preferred
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-900-02110 Supplier Advanced Materials Technology Supplier No. 92812601
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.