Learn More
Alkali Scientific Halo™ Pipettor Starter kit includes 4 Halo™ Pipettors (10µL, 20µL, 200µL, 1250µL) and a a 6 place carousel stand, Free Personalized Engraving (40 custom characters to be spread out over 2 lines of 20)
SDP
Catalog No. 5000662234
Encompass_Preferred
Halo™ Pipettor Starter Kit takes a modern approach to liquid handling with an ergonomics focused design that improves comfort and precision during daily laboratory work. Halo Pipettors are among the lightest pipettors available, helping reduce repetitive stress injuries and minimizing shoulder, arm, and hand fatigue during extended pipetting sessions. Reduced actuation forces and balanced construction allow the pipettor to feel natural and stable in hand. A four digit micrometer display enables precise volume selection, while the click based adjustment wheel with DriftGuard™ protection maintains accuracy during repeated use. The recessed ejector button and cushioned, shock absorbing stop improve tip ejection ergonomics and reduce thumb strain. Halo pipettors are available in multiple volume ranges, including an extended capacity 1250 µL model designed to maximize compatibility with long pipette tips and demanding lab workflows.
Catalog No. 50-006-62234 Supplier Alkali Scientific Supplier No. P9432
Halo™ Pipettor Starter Kit takes a modern approach to liquid handling with an ergonomics focused design that improves comfort and precision during daily laboratory work. Halo Pipettors are among the lightest pipettors available, helping reduce repetitive stress injuries and minimizing shoulder, arm, and hand fatigue during extended pipetting sessions. Reduced actuation forces and balanced construction allow the pipettor to feel natural and stable in hand. A four digit micrometer display enables precise volume selection, while the click based adjustment wheel with DriftGuard™ protection maintains accuracy during repeated use. The recessed ejector button and cushioned, shock absorbing stop improve tip ejection ergonomics and reduce thumb strain. Halo pipettors are available in multiple volume ranges, including an extended capacity 1250 µL model designed to maximize compatibility with long pipette tips and demanding lab workflows.