Learn More
CleanSpace™ HALO Power System NIOSH (Exc Mask)
Catalog No. 17800646 Shop All Cleanspace Technology Products
Change view
Click to view available options
HALO Power System NIOSH (Exc Mask)
Catalog No.
17-800-646 HALO Power System NIOSH (Exc Mask)
1 options
This item is not returnable. View return policy
Catalog No. 17-800-646 Supplier CleanSpace™ Supplier No. CS3020
You must be signed in and have a business account to purchase online.
This item is not returnable. View return policy
Battery-powered, positive pressure air-purifying respirators suitable for healthcare, laboratory, pharmaceutical, and emergency response
  • IP Rating 66 – resistant to dust and liquid ingress
  • Offers protection in applications with particulate contaminates and biohazards
  • Suitable for decontamination procedures
  • Light weight (350 g),compact and comfortable
  • Long operating time: up to 9 hours and quick battery recharging: < 2hours
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Easy and accurate flow capability check
Specifications
Product Type PAPR Assembly, No Headtop
For Use With (Equipment) CS3003, CS3004, CS3005, CS3006,CS3007
Includes Respirator, Neck Support, HEPA filter, Charger
Model CS3020