Learn More
Advanced Materials Technology HALO 90 Å RP-Amide, 2 µm, 2.1 x 30 mm
Supplier:  Advanced Materials Technology 91812307
Encompass_Preferred
HALO RP-Amide columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. NC1638129