Learn More
Alkali Scientific Halo™ Single Channel Pipettor, 200-1250µL
SDP
Catalog No. 502335741
Encompass_Preferred
Halo™ Single Channel Pipettors are exceptionally lightweight, universal fit, adjustable volume, manual pipettes with a four digit micrometer for precise accuracy. They feature a click-stop adjustment wheel with Driftguard™ protection to maintain accurate volume dispensing.
Catalog No. 50-233-5741 Supplier Alkali Scientific Supplier No. P93401MXL
Halo™ Single Channel Pipettors are exceptionally lightweight, universal fit, adjustable volume, manual pipettes with a four digit micrometer for precise accuracy. They feature a click-stop adjustment wheel with Driftguard™ protection to maintain accurate volume dispensing.