Learn More
Alkali Scientific Halo™ Single Channel Pipettor, 2-20µL
SDP
Catalog No. 502335883
Encompass_Preferred
Halo™ Single Channel Pipettors are exceptionally lightweight, universal fit, adjustable volume, manual pipettes with a four digit micrometer for precise accuracy. They feature a click-stop adjustment wheel with Driftguard™ protection to maintain accurate volume dispensing.
Catalog No. 50-233-5883 Supplier Alkali Scientific Supplier No. P934020
Halo™ Single Channel Pipettors are exceptionally lightweight, universal fit, adjustable volume, manual pipettes with a four digit micrometer for precise accuracy. They feature a click-stop adjustment wheel with Driftguard™ protection to maintain accurate volume dispensing.