Learn More
Alkali Scientific Halo™ Single Channel Pipettor, 1 10mL
Supplier: Alkali Scientific P934010M
Halo™ Single Channel Pipettes are exceptionally light , universal fit adjustable volume manual pipettes designed for precise and reliable laboratory liquid handling. Each pipette features a clear four digit micrometer display that allows accurate volume setting and easy reading during routine pipetting tasks. Halo™ manual pipettes are available in ten overlapping volume ranges, enabling researchers to cover a wide spectrum of applications from as low as 0.2 µL up to 10 µL with dependable precision. The variable volume design includes a smooth click stop adjustment wheel that allows fine control while helping prevent accidental changes during use. Integrated Driftguard™ protection helps maintain consistent dispensing accuracy over time. These pipettes are engineered for ergonomic comfort during extended use and are built for durability in demanding laboratory environments. Halo™ manual pipettes are backed by a 3 year manufacturer warranty for added confidence an