Learn More
Alkali Scientific Halo™ Single Channel Pipettor, 10 100µL
SDP
Catalog No. 5000663282
Encompass_Preferred
Halo™ Single Channel Pipettes are exceptionally light , universal fit adjustable volume manual pipettes designed for precise and comfortable laboratory liquid handling. Each pipette features a clear four digit micrometer display that allows users to set and verify volumes with excellent accuracy and repeatability. Halo™ manual pipettes are available in ten overlapping volume ranges, providing coverage for a wide spectrum of applications from as low as 0.2 µL up to 10 µL. The variable volume design includes a smooth click stop adjustment wheel with integrated Driftguard™ protection, helping maintain consistent volume settings and preventing accidental changes during use. Engineered for reliability and everyday laboratory workflows, Halo™ manual pipettes combine ergonomic handling, dependable precision, and durable construction, and are backed by a three year manufacturer warranty for long term performance and confidence.
Catalog No. 50-006-63282 Supplier Alkali Scientific Supplier No. P9340100
Halo™ Single Channel Pipettes are exceptionally light , universal fit adjustable volume manual pipettes designed for precise and comfortable laboratory liquid handling. Each pipette features a clear four digit micrometer display that allows users to set and verify volumes with excellent accuracy and repeatability. Halo™ manual pipettes are available in ten overlapping volume ranges, providing coverage for a wide spectrum of applications from as low as 0.2 µL up to 10 µL. The variable volume design includes a smooth click stop adjustment wheel with integrated Driftguard™ protection, helping maintain consistent volume settings and preventing accidental changes during use. Engineered for reliability and everyday laboratory workflows, Halo™ manual pipettes combine ergonomic handling, dependable precision, and durable construction, and are backed by a three year manufacturer warranty for long term performance and confidence.