Learn More
Proteintech Group Inc Halo-Trap Agarose beads
SDP
Catalog No. 502258558 Shop All Proteintech Group Inc Products
Encompass
Halo-Trap Agarose are affinity beads for IP of Halo-tag proteins. It comprises a Halo-tag Nanobody /VHH conjugated to agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8558 Supplier Proteintech Group Inc Supplier No. OTA20
Halo-Trap Agarose are affinity beads for IP of Halo-tag proteins. It comprises a Halo-tag Nanobody /VHH conjugated to agarose beads.