Learn More
Advanced Materials Technology HALO 90 Å ES-CN, 2.7 µm, 3.0 x 150 mm
Supplier:  Advanced Materials Technology 92813704
Encompass_Preferred
HALO ES-CN columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å ES-CN columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-148-174