Learn More
Advanced Materials Technology HALO 90 Å ES-CN, 2.7 µm, 2.1 x 5 mm, Guard Column 3pk
Catalog No. 50148180
Encompass_Preferred
HALO ES-CN columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å ES-CN columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-148-180 Supplier Advanced Materials Technology Supplier No. 92812104
HALO ES-CN columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å ES-CN columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.