Learn More
ERA WATERS CORP HALOACETIC ACID
SDP
Catalog No. NC9021033
Encompass
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Includes all the analytes below at 5-50 ug/L.
Catalog No. NC9021033 Supplier ERA WATERS CORP Supplier No. 684
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Includes all the analytes below at 5-50 ug/L.