Learn More
Haloacetic Acids Standard, EPA Method 552, 1 mL
Specifications
Specifications
| Chemical Name or Material | Haloacetic Acid Mixture |
| CAS | 1634-04-4, 79-11-8, 79-43-6, 76-03-9, 120-83-2, 79-08-3 |
| CAS Max % | 98.12, 0.14, 0.14, 0.14, 0.14, 0.14 |
| Analyte or Component Names | methyl tert-butyl ether,chloroacetic acid,dichloroacetic acid |
| Element | Methyl Tert-butyl Ether, Chloroacetic Acid, Dichloroacetic Acid |
| Solvent or Matrix | MTBE |
| Quantity | 1 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.