Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haloacetic Acids Mixture (Acids or Methyl Acids) (For EPA 552.2)

Catalog No. USPHM5523A1
Encompass
Change view
Click to view available options
Quantity:
1 mL (Acids)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPHM5523A1 1 mL (Acids)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPHM5523A1 Supplier Agilent Technologies Supplier No. PHM5523A1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Haloacetic Acids Mixture (Acids or Methyl Acids)
Packaging Ampule
Quantity 1 mL (Acids)
Grade EPA
Concentration or Composition (by Analyte or Components) Each component in Methyl tert-Butyl Ether:
Chloroacetic Acid    •600μg/mL
Chlorodibromoacetic Acid    •1000μg/mL
Dichloroacetic Acid    •600μg/mL
Trichloroacetic Acid    •200μg/mL
Bromoacetic Acid    •400μg/mL
Bromochloroacetic Acid    •400μg/mL
Bromodichloroacetic Acid    •400μg/mL
Dibromoacetic Acid    •200μg/mL
Tribromoacetic Acid    •2000μg/mL
Dalapon    •400μg/mL
2,3-Dibromopropionic    •1000μg/mL
Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.