Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC 700904
Encompass
Resistant to rifampin [rifampicin; rifamycin AMP], penicillin, clindamycin, erythromycin, tetracycline, and chloramphenicol. Representative strain of the Pneumococcal Molecular Epidemiology Network (PMEN).
Catalog No. 50-237-9845