Learn More
ATCC Legionella pneumophila ;subsp. pneumophila
Catalog No. 502381467
Encompass
Legionella pneumophila subsp. pneumophila strain Togus-1 was isolated from a human lung. This strain is nutritionally fastidious and can be inhibited by media of poor caliber, so strict attention should be paid to the quality of the ingredients.
Catalog No. 50-238-1467 Supplier ATCC Supplier No. 33154
Legionella pneumophila subsp. pneumophila strain Togus-1 was isolated from a human lung. This strain is nutritionally fastidious and can be inhibited by media of poor caliber, so strict attention should be paid to the quality of the ingredients.