Learn More
ATCC Finegoldia magna; Strain WAL2508
Supplier:  ATCC 29328
Encompass
Finegoldia magna strain WAL2508 is a bacterial anaerobe that was isolated from an abdominal wound. This strain has applications as a quality control strain for MicroScan™ products.
Catalog No. 50-237-9721