Learn More
ATCC Halobacterium salinarum
Catalog No. 502379851
Encompass
Halobacterium salinarum strain NRC-1 is a whole-genome sequenced archaeon that contains the megaplasmid or minichromosome pNRC100. This strain is a halophile.
Catalog No. 50-237-9851 Supplier ATCC Supplier No. 700922
Halobacterium salinarum strain NRC-1 is a whole-genome sequenced archaeon that contains the megaplasmid or minichromosome pNRC100. This strain is a halophile.