Learn More
Cayman Chemical Halobetasol PropIonate 25mg
SDP
Catalog No. 502994798
Encompass_Preferred
A synthetic glucocorticoid; inhibits dermatitis induced by ultraviolet light in guinea pigs (ED50 = 1.45 UG/animal) and by oxazolone in mice (ED50 = 0.8 UG/animal); has anti-inflammatory effects on croton oil-induced EAr edema in rats (ED50s = 10 UG/ml and 0.5 UG/ear) and mice (ED50s = 1.2 and <0.32 UG/ear after 6 and 24 hours of treatment, respectively)
Catalog No. 50-299-4798 Supplier Cayman Chemical Supplier No. 2378125 MG
A synthetic glucocorticoid; inhibits dermatitis induced by ultraviolet light in guinea pigs (ED50 = 1.45 UG/animal) and by oxazolone in mice (ED50 = 0.8 UG/animal); has anti-inflammatory effects on croton oil-induced EAr edema in rats (ED50s = 10 UG/ml and 0.5 UG/ear) and mice (ED50s = 1.2 and <0.32 UG/ear after 6 and 24 hours of treatment, respectively)