Learn More
Cayman Chemical Halobetasol Propionate, Purity: ≥95%, CAS: 66852-54-8, 50 mg
SDP
Catalog No. 502994561
Encompass_Preferred
A synthetic glucocorticoid; inhibits dermatitis induced by ultraviolet light in guinea pigs (ED50 = 1.45 µg/animal) and by oxazolone in mice (ED50 = 0.8 µg/animal); has anti-inflammatory effects on croton oil-induced ear edema in rats (ED50s = 10 µg/ml and 0.5 µg/ear) and mice (ED50s = 1.2 and <0.32 µg/ear after 6 and 24 hours of treatment, respectively)
Catalog No. 50-299-4561 Supplier Cayman Chemical Supplier No. 2378150 MG
A synthetic glucocorticoid; inhibits dermatitis induced by ultraviolet light in guinea pigs (ED50 = 1.45 µg/animal) and by oxazolone in mice (ED50 = 0.8 µg/animal); has anti-inflammatory effects on croton oil-induced ear edema in rats (ED50s = 10 µg/ml and 0.5 µg/ear) and mice (ED50s = 1.2 and <0.32 µg/ear after 6 and 24 hours of treatment, respectively)