Learn More
ATCC Streptococcus pneumoniae; Strain 178
Supplier:  ATCC BAA343
Encompass
Streptococcus pneumoniae strain 178 [Poland 23F-16] is a drug-resistant bacterium that was isolated in Poland from the broncheoalveolar lavage from a girl with a lower respiratory tract infection. This strain is serotype 23F and is a representative strain of the Poland23F-16 pneumococcal clone as confirmed by the Pneumococcal Molecular Epidemiology Network (PMEN).
Catalog No. 50-238-0923
 Restricted Item