Learn More
Medchemexpress LLC Halofantrine hydrochloride | 36167-63-2 | 99.9% | C26H31Cl3F3NO | 1 MG
SDP
Catalog No. 5000432316
Encompass_Preferred
Halofantrine hydrochloride is a blocker that delays the delayed rectifier potassium current by inhibiting human ERG channels, and it is a potent antimalarial agent. It inhibits the Cap1-dependent oxidative stress response of Candida albicans, suppresses ROS responses, and enhances the antifungal activity of oxidative damage agents.
  • Exhibits antifungal activity in the *Galleria mellonella* model.
  • Shows antimalarial activity against *Plasmodium* strains both *in vitro* and in animal models.
  • Can be used in studies related to invasive candidiasis, falciparum malaria, and vivax malaria.
Catalog No. 50-004-32316 Supplier Medchemexpress LLC Supplier No. HYA0148A1MG
Halofantrine hydrochloride is a blocker that delays the delayed rectifier potassium current by inhibiting human ERG channels, and it is a potent antimalarial agent. It inhibits the Cap1-dependent oxidative stress response of Candida albicans, suppresses ROS responses, and enhances the antifungal activity of oxidative damage agents.
  • Exhibits antifungal activity in the *Galleria mellonella* model.
  • Shows antimalarial activity against *Plasmodium* strains both *in vitro* and in animal models.
  • Can be used in studies related to invasive candidiasis, falciparum malaria, and vivax malaria.