Learn More
AdipoGen Halofenozide
SDP
Catalog No. 502560741
Encompass_Preferred
Chemical. CAS 112226-61-6. Formula C18H19ClN2O2. MW 330.8. Synthetic. Dibenzoylhydrazine based nonsteroidal insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.
Catalog No. 50-256-0741 Supplier AdipoGen Supplier No. CDXH0097M250
Chemical. CAS 112226-61-6. Formula C18H19ClN2O2. MW 330.8. Synthetic. Dibenzoylhydrazine based nonsteroidal insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.