Learn More
Cayman Chemical HalofntrInhydrochlorIde 10mg
SDP
Catalog No. 502988359
Encompass_Preferred
An antimalarial agent; active against chloroquine-sensitive and chloroquine-resistant strains of P. falciparum (IC50s = 1.5-2.5 and 1.3-3.9 UG/L, respectively); reduces parasitemia in a mouse model of P. berghei infection (CD50 = 15 mg/kg); reduces parasitemia in an Aotus monkey model of P. falciparum infection (CD50 = 58.3 mg/kg)
Catalog No. 50-298-8359 Supplier Cayman Chemical Supplier No. 3096210 MG
An antimalarial agent; active against chloroquine-sensitive and chloroquine-resistant strains of P. falciparum (IC50s = 1.5-2.5 and 1.3-3.9 UG/L, respectively); reduces parasitemia in a mouse model of P. berghei infection (CD50 = 15 mg/kg); reduces parasitemia in an Aotus monkey model of P. falciparum infection (CD50 = 58.3 mg/kg)