Learn More
TARGETMOL CHEMICALS INC HALOFUGINONE 10MG
SDP
Catalog No. 502576433
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Halofuginone (RU-19110) the competitive inhibitor of prolyl-tRNA synthetase(Ki=18.3 nM) could also down-regulate Smad3 and blocked TGF-(beta) signaling at 10 ng/ml in the mammal. purity: 97%
Catalog No. 50-257-6433 Supplier TARGETMOL CHEMICALS INC Supplier No. T685610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Halofuginone (RU-19110) the competitive inhibitor of prolyl-tRNA synthetase(Ki=18.3 nM) could also down-regulate Smad3 and blocked TGF-(beta) signaling at 10 ng/ml in the mammal. purity: 97%