Learn More
Selleck Chemical LLC Halofuginone-5mg
SDP
Catalog No. 502390028
Encompass_Preferred
Halofuginone (RU-19110) is the competitively inhibitor of prolyl-tRNA synthetase with Ki of 18.3 nM.It could also down-regulate Smad3 and blocked TGF-β signaling at 10 ng/ml in mammal.
Catalog No. 50-239-0028 Supplier Selleck Chemical LLC Supplier No. S81445MG
Halofuginone (RU-19110) is the competitively inhibitor of prolyl-tRNA synthetase with Ki of 18.3 nM.It could also down-regulate Smad3 and blocked TGF-β signaling at 10 ng/ml in mammal.