Learn More
Selleck Chemical LLC Halofuginone hydrobromide E2709-25MG
SDP
Catalog No. 5000039636
Encompass_Preferred
Halofuginone (RU-19110) hydrobromid a febrifugine derivative is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18 3 nM Halofuginone hydrobromid is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibition of TGF- activity
Catalog No. 50-000-39636 Supplier Selleck Chemical LLC Supplier No. E270925MG
Halofuginone (RU-19110) hydrobromid a febrifugine derivative is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18 3 nM Halofuginone hydrobromid is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibition of TGF- activity