Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | 495.59 | 100 MG
SDP
Catalog No. 5000416370
Encompass_Preferred
Halofuginone hydrobromide (RU-19110 hydrobromide) is a competitive prolyl-tRNA synthetase inhibitor. This Febrifugine derivative acts as a specific inhibitor of type-I collagen synthesis and helps in attenuating osteoarthritis by inhibiting TGF-β activity. It also functions as a potent pulmonary vasodilator through the activation of Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Acts as a competitive prolyl-tRNA synthetase inhibitor.
  • Inhibits type-I collagen synthesis.
  • Attenuates osteoarthritis by inhibiting TGF-β activity.
  • Functions as a potent pulmonary vasodilator.
  • Activates Kv channels.
  • Blocks voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Possesses anti-malaria effects.
  • Demonstrates anti-inflammatory properties.
  • Exhibits anti-cancer and anti-fibrosis effects.
Catalog No. 50-004-16370 Supplier Medchemexpress LLC Supplier No. HYN1584A100MG
Halofuginone hydrobromide (RU-19110 hydrobromide) is a competitive prolyl-tRNA synthetase inhibitor. This Febrifugine derivative acts as a specific inhibitor of type-I collagen synthesis and helps in attenuating osteoarthritis by inhibiting TGF-β activity. It also functions as a potent pulmonary vasodilator through the activation of Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Acts as a competitive prolyl-tRNA synthetase inhibitor.
  • Inhibits type-I collagen synthesis.
  • Attenuates osteoarthritis by inhibiting TGF-β activity.
  • Functions as a potent pulmonary vasodilator.
  • Activates Kv channels.
  • Blocks voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Possesses anti-malaria effects.
  • Demonstrates anti-inflammatory properties.
  • Exhibits anti-cancer and anti-fibrosis effects.