Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | 495.59 | 25 MG
SDP
Catalog No. 5000389165
Encompass_Preferred
Halofuginone (hydrobromide) (Standard) is the analytical standard of Halofuginone (hydrobromide). It is a Febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. This product is intended for research and analytical applications.
  • Analytical standard for Halofuginone (hydrobromide)
  • Competitive prolyl-tRNA synthetase inhibitor (Ki of 18.3 nM)
  • Specific inhibitor of type-I collagen synthesis
  • Attenuates osteoarthritis (OA) by inhibiting TGF-β activity
  • Potent pulmonary vasodilator (activates Kv channels and blocks voltage-gated, receptor-operated and store-operated Ca2+ channels)
  • Anti-malaria, anti-inflammatory, anti-cancer, and anti-fibrosis effects
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-89165 Supplier Medchemexpress LLC Supplier No. 25MGHYN1584AR
Halofuginone (hydrobromide) (Standard) is the analytical standard of Halofuginone (hydrobromide). It is a Febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. This product is intended for research and analytical applications.
  • Analytical standard for Halofuginone (hydrobromide)
  • Competitive prolyl-tRNA synthetase inhibitor (Ki of 18.3 nM)
  • Specific inhibitor of type-I collagen synthesis
  • Attenuates osteoarthritis (OA) by inhibiting TGF-β activity
  • Potent pulmonary vasodilator (activates Kv channels and blocks voltage-gated, receptor-operated and store-operated Ca2+ channels)
  • Anti-malaria, anti-inflammatory, anti-cancer, and anti-fibrosis effects
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS