Learn More
Medchemexpress LLC Halofuginone (hydrobromide) | 64924-67-0 | 495.59 | 25 MG
SDP
Catalog No. 5000389998
Encompass_Preferred
Halofuginone hydrobromide, a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor. It specifically inhibits type-I collagen synthesis and helps reduce osteoarthritis by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking various Ca2+ channels. This compound exhibits a range of beneficial effects.
  • Competitively inhibits prolyl-tRNA synthetase
  • Inhibits type-I collagen synthesis
  • Attenuates osteoarthritis by inhibiting TGF-β activity
  • Potent pulmonary vasodilator
  • Activates Kv channels and blocks Ca2+ channels
  • Exhibits anti-malaria effects
  • Provides anti-inflammatory effects
  • Shows anti-cancer activity
  • Offers anti-fibrosis effects
  • Decreases NRF2 protein levels in tumors
Catalog No. 50-003-89998 Supplier Medchemexpress LLC Supplier No. 25MGHYN1584A
Halofuginone hydrobromide, a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor. It specifically inhibits type-I collagen synthesis and helps reduce osteoarthritis by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking various Ca2+ channels. This compound exhibits a range of beneficial effects.
  • Competitively inhibits prolyl-tRNA synthetase
  • Inhibits type-I collagen synthesis
  • Attenuates osteoarthritis by inhibiting TGF-β activity
  • Potent pulmonary vasodilator
  • Activates Kv channels and blocks Ca2+ channels
  • Exhibits anti-malaria effects
  • Provides anti-inflammatory effects
  • Shows anti-cancer activity
  • Offers anti-fibrosis effects
  • Decreases NRF2 protein levels in tumors