Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | 495.59 | 50 MG
SDP
Catalog No. 5000415606
Encompass_Preferred
Halofuginone hydrobromide (RU-19110) is a Febrifugine derivative that acts as a competitive prolyl-tRNA synthetase inhibitor. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. This compound exhibits various biological effects, including anti-malarial, anti-inflammatory, anti-cancer, and anti-fibrosis activities. It also functions as a potent pulmonary vasodilator.
  • Competitive prolyl-tRNA synthetase inhibitor.
  • Inhibits type-I collagen synthesis.
  • Attenuates osteoarthritis by inhibiting TGF-β activity.
  • Potent pulmonary vasodilator, activating Kv channels and blocking Ca2+ channels.
  • Exhibits anti-malarial, anti-inflammatory, anti-cancer, and anti-fibrosis effects.
  • Decreases NRF2 protein levels in tumors.
  • Partially reverses established pulmonary hypertension.
Catalog No. 50-004-15606 Supplier Medchemexpress LLC Supplier No. HYN1584A50MG
Halofuginone hydrobromide (RU-19110) is a Febrifugine derivative that acts as a competitive prolyl-tRNA synthetase inhibitor. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. This compound exhibits various biological effects, including anti-malarial, anti-inflammatory, anti-cancer, and anti-fibrosis activities. It also functions as a potent pulmonary vasodilator.
  • Competitive prolyl-tRNA synthetase inhibitor.
  • Inhibits type-I collagen synthesis.
  • Attenuates osteoarthritis by inhibiting TGF-β activity.
  • Potent pulmonary vasodilator, activating Kv channels and blocking Ca2+ channels.
  • Exhibits anti-malarial, anti-inflammatory, anti-cancer, and anti-fibrosis effects.
  • Decreases NRF2 protein levels in tumors.
  • Partially reverses established pulmonary hypertension.