Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | 495.59 | 50 MG
SDP
Catalog No. 5000416371
Encompass_Preferred
Halofuginone hydrobromide (Standard) is an analytical standard derived from Febrifugine, functioning as a competitive prolyl-tRNA synthetase inhibitor. It also acts as a specific inhibitor of type-I collagen synthesis, reducing osteoarthritis symptoms by suppressing TGF-β activity. Additionally, it is a potent pulmonary vasodilator, enhancing Kv channels and blocking various calcium channels, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Attenuates osteoarthritis
  • Activates Kv channels
  • Blocks voltage-gated, receptor-operated, and store-operated calcium channels
  • Has anti-malaria effects
  • Has anti-inflammatory effects
  • Has anti-cancer effects
  • Has anti-fibrosis effects
  • Used in HPLC, GC, and MS research
Catalog No. 50-004-16371 Supplier Medchemexpress LLC Supplier No. HYN1584AR50MG
Halofuginone hydrobromide (Standard) is an analytical standard derived from Febrifugine, functioning as a competitive prolyl-tRNA synthetase inhibitor. It also acts as a specific inhibitor of type-I collagen synthesis, reducing osteoarthritis symptoms by suppressing TGF-β activity. Additionally, it is a potent pulmonary vasodilator, enhancing Kv channels and blocking various calcium channels, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Attenuates osteoarthritis
  • Activates Kv channels
  • Blocks voltage-gated, receptor-operated, and store-operated calcium channels
  • Has anti-malaria effects
  • Has anti-inflammatory effects
  • Has anti-cancer effects
  • Has anti-fibrosis effects
  • Used in HPLC, GC, and MS research