Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | 495.59 | 500 MG
SDP
Catalog No. 5000416488
Encompass_Preferred
Halofuginone hydrobromide is a Febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Competitive prolyl-tRNA synthetase inhibitor (Ki of 18.3 nM).
  • Specific inhibitor of type-I collagen synthesis.
  • Attenuates osteoarthritis (OA) by inhibition of TGF-β activity.
  • Potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Has anti-malaria, anti-inflammatory, anti-cancer, and anti-fibrosis effects.
Catalog No. 50-004-16488 Supplier Medchemexpress LLC Supplier No. HYN1584A500MG
Halofuginone hydrobromide is a Febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Competitive prolyl-tRNA synthetase inhibitor (Ki of 18.3 nM).
  • Specific inhibitor of type-I collagen synthesis.
  • Attenuates osteoarthritis (OA) by inhibition of TGF-β activity.
  • Potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Has anti-malaria, anti-inflammatory, anti-cancer, and anti-fibrosis effects.