Learn More
Medchemexpress LLC Halofuginone (hydrobromide) | 64924-67-0 | MFCD00946455 | 100.0% | 495.59 g/mol | C16H18Br2ClN3O3 | 10 MG
SDP
Catalog No. 5000295701
Encompass_Preferred
Halofuginone hydrobromide is the hydrobromide salt of halofuginone, a febrifugine-derived small molecule that competitively inhibits prolyl-tRNA synthetase and is supplied for research applications studying fibrosis, inflammation, malaria, and cancer. It is provided as a white to off-white solid with high purity and defined storage and solvent recommendations.
  • Hydrobromide salt for improved handling and solubility.
  • Competitive prolyl-tRNA synthetase inhibitor used in mechanistic research.
  • High purity suitable for research applications.
  • White to off-white solid, soluble in DMSO (≈50 mg/mL) and water (≈2.6 mg/mL).
  • Store sealed and away from moisture; in solution keep at -80°C for long-term stability.
  • Applicable for preclinical studies of fibrosis, inflammation, malaria, and cancer pathways.
Catalog No. 50-002-95701 Supplier Medchemexpress LLC Supplier No. HYN1584A10MG
Halofuginone hydrobromide is the hydrobromide salt of halofuginone, a febrifugine-derived small molecule that competitively inhibits prolyl-tRNA synthetase and is supplied for research applications studying fibrosis, inflammation, malaria, and cancer. It is provided as a white to off-white solid with high purity and defined storage and solvent recommendations.
  • Hydrobromide salt for improved handling and solubility.
  • Competitive prolyl-tRNA synthetase inhibitor used in mechanistic research.
  • High purity suitable for research applications.
  • White to off-white solid, soluble in DMSO (≈50 mg/mL) and water (≈2.6 mg/mL).
  • Store sealed and away from moisture; in solution keep at -80°C for long-term stability.
  • Applicable for preclinical studies of fibrosis, inflammation, malaria, and cancer pathways.