Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | MFCD00946455 | 100.0% | 495.59 g·mol⁻¹ | C16H18Br2ClN3O3 | 10 MG
SDP
Catalog No. 5000295702
Encompass_Preferred
Halofuginone hydrobromide is a febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor used as a research standard for studies of collagen synthesis, TGF-β signaling, fibrosis, cancer biology, and anti-parasitic activity.
  • Competitive prolyl-tRNA synthetase inhibitor with reported Ki ≈ 18.3 nM.
  • Inhibits type-I collagen synthesis, useful for fibrosis and TGF-β studies.
  • High reported purity (99.99%), suitable as a reference standard.
  • Characterized molecular weight (495.59 g·mol⁻¹) and CAS number (64924-67-0).
  • Available in small mg quantities for in vitro and pharmacological assays.
Catalog No. 50-002-95702 Supplier Medchemexpress LLC Supplier No. HYN1584AR10MG
Halofuginone hydrobromide is a febrifugine derivative and a competitive prolyl-tRNA synthetase inhibitor used as a research standard for studies of collagen synthesis, TGF-β signaling, fibrosis, cancer biology, and anti-parasitic activity.
  • Competitive prolyl-tRNA synthetase inhibitor with reported Ki ≈ 18.3 nM.
  • Inhibits type-I collagen synthesis, useful for fibrosis and TGF-β studies.
  • High reported purity (99.99%), suitable as a reference standard.
  • Characterized molecular weight (495.59 g·mol⁻¹) and CAS number (64924-67-0).
  • Available in small mg quantities for in vitro and pharmacological assays.