Learn More
Medchemexpress LLC Halofuginone hydrobromide | 64924-67-0 | MFCD00946455 | 495.59 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN1584AR100MG
Encompass_Preferred
Halofuginone hydrobromide (Standard) is the analytical standard of Halofuginone (hydrobromide) and is intended for research and analytical applications. It is a Febrifugine derivative, acting as a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Specific inhibitor of type-I collagen synthesis.
  • Attenuates osteoarthritis (OA) by inhibiting TGF-β activity.
  • Potent pulmonary vasodilator through activation of Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Possesses anti-malaria, anti-inflammatory, anti-cancer, and anti-fibrosis effects.
Catalog No. 50-004-15605