Learn More
Medchemexpress LLC Halofuginone hydrochloride | 1217623-74-9 | 451.14 | 1 MG
SDP
Catalog No. 5000414855
Encompass_Preferred
Halofuginone hydrobromid, a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Exhibits anti-malaria effects.
  • Exhibits anti-inflammatory effects.
  • Exhibits anti-cancer effects.
  • Exhibits anti-fibrosis effects.
Catalog No. 50-004-14855 Supplier Medchemexpress LLC Supplier No. HYN1584B1MG
Halofuginone hydrobromid, a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM. It is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibiting TGF-β activity. It also acts as a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated, and store-operated Ca2+ channels.
  • Exhibits anti-malaria effects.
  • Exhibits anti-inflammatory effects.
  • Exhibits anti-cancer effects.
  • Exhibits anti-fibrosis effects.