Learn More
Bulbman Inc 20W 120V G8 BIPIN HALOGEN BULB
SDP
Catalog No. NC0302291
Encompass_Preferred
20 watt, 120 volt, GY8 base, 2900K color temperature, clear, 1.33" short T4 halogen bulb with an average life of 2000 hours.
Catalog No. NC0302291 Supplier Bulbman Inc Supplier No. 132019
20 watt, 120 volt, GY8 base, 2900K color temperature, clear, 1.33" short T4 halogen bulb with an average life of 2000 hours.